Skip to Content
Merck
All Photos(1)

Key Documents

V800157

Sigma-Aldrich

Diethyl ether

≥99.5%, anhydrous, contains 5 ppm BHT as stabilizer, Vetec

Synonym(s):

Et2O, Ethoxyethane, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
anhydrous
bp:
34.6 °C (lit.)

Product Name

Diethyl ether, anhydrous, contains 5 ppm BHT as stabilizer, ≥99.5%

grade

anhydrous

vapor density

2.6 (vs air)

product line

Vetec

Assay

≥99.5%

form

liquid

autoignition temp.

320 °F

contains

5 ppm BHT as stabilizer

expl. lim.

36.5 %

dilution

(for analytical testing)

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

density

0.706 g/mL at 25 °C (lit.)

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan E Camacho Londoño et al.
European heart journal, 36(33), 2257-2266 (2015-06-13)
Pathological cardiac hypertrophy is a major predictor for the development of cardiac diseases. It is associated with chronic neurohumoral stimulation and with altered cardiac Ca(2+) signalling in cardiomyocytes. TRPC proteins form agonist-induced cation channels, but their functional role for Ca(2+)
Faysal Benaskar et al.
ChemSusChem, 6(2), 353-366 (2012-11-30)
A μ(2)-process in the Ullmann-type C-O coupling of potassium phenolate and 4-chloropyridine was successfully performed in a combined microwave (MW) and microflow process. Selective MW absorption in a micro-fixed-bed reactor (μ-FBR) by using a supported Cu nanocatalyst resulted in an
Ether-directed ortho-C-H olefination with a palladium(II)/monoprotected amino acid catalyst.
Gang Li et al.
Angewandte Chemie (International ed. in English), 52(4), 1245-1247 (2012-12-15)
Tatsuya Yoshino et al.
Organic letters, 14(16), 4290-4292 (2012-08-09)
A highly efficient total synthesis of the 11-membered cyclic aspercyclides A (1) and B (2) has been achieved by chemo- and regioselective intramolecular oxidative C-O bond formation from differently substituted diphenols.
Alessandra Forni et al.
Journal of molecular graphics & modelling, 38, 31-39 (2012-10-23)
The solvent effect on the I⋯O halogen bonding in complexes of iodobenzene derivatives with formaldehyde has been investigated by systematically varying the substituents on the iodobenzene ring. Calculations have been performed at MP2 and DFT levels of theory, using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service