Skip to Content
Merck
All Photos(1)

Key Documents

V800140

Sigma-Aldrich

Cyclohexanone

LR, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14
Beilstein:
385735
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
grade:
LR
bp:
155 °C (lit.)
vapor pressure:
3.4 mmHg ( 20 °C)

grade

LR

vapor density

3.4 (vs air)

vapor pressure

3.4 mmHg ( 20 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

788 °F

expl. lim.

1.1 %, 100 °F
9.4 %

dilution

(for analytical testing)

refractive index

n20/D 1.450 (lit.)

pH

~7 (20 °C, 70 g/L)

bp

155 °C (lit.)

mp

−47 °C (lit.)

density

0.947 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Kleinschmidt et al.
Journal of colloid and interface science, 559, 76-87 (2019-10-15)
Exploring and controlling chemical reactions in compartments opens new platforms for designing bioinspired catalysts and energy-autonomous systems. Aqueous polymer networks or hydrogels serve as a perfect model for biological tissues, allowing systematic investigations of chemical transformations in compartments. Herein, we
Alakananda Hajra et al.
Organic letters, 14(21), 5488-5491 (2012-10-18)
Dehydrogenative aromatization of cyclohexanone imines to arylamines has been achieved using a palladium catalyst under aerobic conditions. The reaction is applicable to a variety of imines that are either preformed or generated in situ from cyclohexanone derivatives and aryl or
Brahm J Yachnin et al.
Journal of the American Chemical Society, 134(18), 7788-7795 (2012-04-18)
The Baeyer-Villiger monooxygenases (BVMOs) are a family of bacterial flavoproteins that catalyze the synthetically useful Baeyer-Villiger oxidation reaction. This involves the conversion of ketones into esters or cyclic ketones into lactones by introducing an oxygen atom adjacent to the carbonyl
Application of fragment screening and merging to the discovery of inhibitors of the Mycobacterium tuberculosis cytochrome P450 CYP121.
Sean A Hudson et al.
Angewandte Chemie (International ed. in English), 51(37), 9311-9316 (2012-08-15)
Viktória Fábos et al.
Chemistry, an Asian journal, 7(11), 2629-2637 (2012-09-07)
Iron (and to a lesser extent manganese) in the wall of a 316 stainless steel (SS) reactor is responsible for the hydrogenation of cyclohexanone to cyclohexanol when using an aqueous formic acid solution under high temperature and pressure water (HTPW)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service