All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
Beilstein:
605632
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
grade:
LR
bp:
132 °C (lit.)
vapor pressure:
11.8 mmHg ( 25 °C)
grade
LR
vapor density
3.86 (vs air)
vapor pressure
11.8 mmHg ( 25 °C)
product line
Vetec™
Assay
≥99%
form
liquid
autoignition temp.
1178 °F
expl. lim.
7.1 %
dilution
(for analytical testing)
refractive index
n20/D 1.524 (lit.)
bp
132 °C (lit.)
mp
−45 °C (lit.)
density
1.106 g/mL at 25 °C (lit.)
SMILES string
Clc1ccccc1
InChI
1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H
InChI key
MVPPADPHJFYWMZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
80.6 °F
Flash Point(C)
27 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Wei-Chao Chen et al.
ACS applied materials & interfaces, 7(8), 4616-4622 (2015-02-11)
Controlled dispersion of single-walled carbon nanotubes (SWCNTs) in common solvents is a challenging issue, especially for the rising need of low cost flexible transparent conducting films (TCFs). Utilizing conductive polymer as surfactant to facilitate SWCNTs solubility is the most successful
Kwanpyo Kim et al.
Nature communications, 5, 4781-4781 (2014-09-03)
One-dimensional defects in graphene have a strong influence on its physical properties, such as electrical charge transport and mechanical strength. With enhanced chemical reactivity, such defects may also allow us to selectively functionalize the material and systematically tune the properties
Kyeongil Hwang et al.
Advanced materials (Deerfield Beach, Fla.), 27(7), 1241-1247 (2015-01-13)
Fully printed perovskite solar cells are demonstrated with slot-die coating, a scalable printing method. A sequential slot-die coating process is developed to produce efficient perovskite solar cells and to be used in a large-scale roll-to-roll printing process. All layers excluding
M Govindarajan et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 88, 37-48 (2011-12-27)
In this work, the vibrational spectral analysis was carried out by using FT-Raman and FT-IR spectroscopy in the range 100-4000 cm(-1) and 400-4000 cm(-1) respectively, for the title molecule. The molecular structure, fundamental vibrational frequencies and intensity of the vibrational
Kerry N McPhedran et al.
Water research, 47(7), 2222-2230 (2013-03-12)
The sorption behaviour of hydrophobic organic compounds (HOCs) 1,2,4,5-tetrachlor-obenzene (TeCB), pentachlorobenzene (PeCB) and hexachlorobenzene (HCB) to Aldrich humic acid (AHA) and municipal wastewater treatment plant (MWTP) influent colloidal organic carbon (COC) was investigated using the gas-stripping technique. Gas stripping assumptions
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service