All Photos(1)
About This Item
Linear Formula:
CaSO4 · 2H2O
CAS Number:
Molecular Weight:
172.17
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
AR
form:
powder
Recommended Products
grade
AR
product line
Vetec™
Assay
≥99%
form
powder
anion traces
chloride (Cl-): ≤0.04%
cation traces
Fe: ≤0.005%
SMILES string
O.O.[Ca++].[O-]S([O-])(=O)=O
InChI
1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2
InChI key
PASHVRUKOFIRIK-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Massimo Peruffo et al.
Physical chemistry chemical physics : PCCP, 15(6), 1956-1965 (2012-12-22)
Dissolution processes at single crystal surfaces often involve the initial formation and expansion of localized, characteristic (faceted) etch-pits at defects, in an otherwise comparatively unreactive surface. Using natural gypsum single crystal as an example, a simple but powerful morphological analysis
Accuracy of dies captured by an intraoral digital impression system using parallel confocal imaging.
So-Yeun Kim et al.
The International journal of prosthodontics, 26(2), 161-163 (2013-03-12)
The purpose of this investigation was to measure the accuracy of digital impressions (DIs) compared to conventional impressions (CIs). Using the iTero system, a master cast was scanned to produce stereolithography dies. As a control group, silicone impressions were taken
Rawaa Ammar et al.
Environmental science and pollution research international, 20(12), 9014-9025 (2013-06-15)
One of the most important sources of solid waste in the Mediterranean Basin ecosystem originated from the phosphate fertilizer industries, which discharge phosphogypsum (PG) directly into aquatic environments or are stacked on stockpiles. The present study investigates metal release from
C M Belfiore et al.
Environmental science and pollution research international, 20(12), 8848-8859 (2013-06-08)
Black crusts are recognized to have been, up to now, one of the major deterioration forms affecting the built heritage in urban areas. Their formation is demonstrated to occur mainly on carbonate building materials, whose interaction with an SO2-loaded atmosphere
Kathryn A Dooley et al.
The Analyst, 138(17), 4838-4848 (2013-06-27)
In situ chemical imaging techniques are being developed to provide information on the spatial distribution of artists' pigments used in polychrome works of art such as paintings. The new methods include reflectance imaging spectroscopy and X-ray fluorescence mapping. Results from
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service