Skip to Content
Merck
All Photos(1)

Key Documents

V800105

Sigma-Aldrich

Calcium hydroxide

LR, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
EC Number:
MDL number:
UNSPSC Code:
12352305
PubChem Substance ID:
grade:
LR
form:
powder

grade

LR

product line

Vetec

Assay

≥95%

form

powder

pH

12.4-12.6 (20 °C)

density

2.24 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.02%
sulfate (SO42-): ≤0.10%

cation traces

heavy metals (as Pb): ≤0.005%

SMILES string

O[Ca]O

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisbet Mailin López González et al.
Bioresource technology, 131, 250-257 (2013-01-29)
Different pre-treatment severities by thermo-alkaline conditions (100°C, Ca(OH)2) on press mud were evaluated for different pre-treatment time and lime loading. COD solubilization and the methane yield enhancement were assessed. The biochemical methane potential was determined in batch assays under mesophilic
Nikte Gómez-Ortíz et al.
ACS applied materials & interfaces, 5(5), 1556-1565 (2013-01-26)
The presence and deteriorating action of microbial biofilms on historic stone buildings have received considerable attention in the past few years. Among microorganisms, fungi are one of the most damaging groups. In the present work, antimicrobial surfaces were prepared using
T J Hilton et al.
Journal of dental research, 92(7 Suppl), 16S-22S (2013-05-22)
This practice-based, randomized clinical trial evaluated and compared the success of direct pulp capping in permanent teeth with MTA (mineral trioxide aggregate) or CaOH (calcium hydroxide). Thirty-five practices in Northwest PRECEDENT were randomized to perform direct pulp caps with either
F H van de Sande et al.
Journal of dental research, 92(7 Suppl), 78S-83S (2013-05-22)
This practice-based retrospective study evaluated the survival of resin composite restorations in posterior teeth, focusing on the influence of potential patient risk factors. In total, 306 posterior composite restorations placed in 44 adult patients were investigated after 10 to 18
Lidan Ye et al.
Bioresource technology, 132, 38-44 (2013-02-13)
Cost-effective production of optically pure lactic acid from lignocellulose sugars is commercially attractive but challenging. Bacillus coagulans C106 was isolated from environment and used to produce l-lactic acid from xylose at 50°C and pH 6.0 in mineral salts medium containing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service