About This Item
Recommended Products
grade
LR
product line
Vetec™
Assay
≥98%
form
solid
pH
5-8 (25 °C, 50 g/L in water)
mp
592 °C (dec.) (lit.)
SMILES string
[Ba++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/Ba.2NO3/c;2*2-1(3)4/q+2;2*-1
InChI key
IWOUKMZUPDVPGQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service