Skip to Content
Merck
All Photos(1)

Key Documents

V800043

Sigma-Aldrich

Ammonium phosphate monobasic

AR, ≥99%

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
AR
form:
crystalline powder

grade

AR

vapor pressure

<0 hPa ( 20 °C)

product line

Vetec

Assay

≥99%

form

crystalline powder

pH

3.8-4.4 (25 °C, 50 g/L)

mp

190 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Young Yang et al.
Clinical chemistry and laboratory medicine, 53(8), 1249-1257 (2015-02-27)
Since the urinary concentration of 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanol (NNAL) is a reliable biomarker of exposure to tobacco smoke, we developed a relatively simple high-throughput chromatographic method to quantify total urinary NNAL concentrations in the general population. The high-throughput analytical method was developed
Nathan K Deed et al.
Applied microbiology and biotechnology, 89(5), 1537-1549 (2011-01-20)
Two deletion mutants expected to be defective in nitrogen catabolite repression (NCR) were constructed in a commercial wine yeast background M2: a ure2 mutant and a dal80 gzf3 double mutant. Wild-type and both mutant strains were fermented in Sauvignon Blanc
Christopher T Mills et al.
The Science of the total environment, 435-436, 363-373 (2012-08-08)
Sacramento Valley (California, USA) soils and sediments have high concentrations of Cr(III) because they are partially derived from ultramafic material. Some Cr(III) is oxidized to more toxic and mobile Cr(VI) by soil Mn oxides. Valley soils typically have neutral to
Catarina Barbosa et al.
International journal of food microbiology, 160(2), 87-93 (2012-11-28)
Sulphur-containing amino acids, cysteine and methionine, are generally found in very low concentrations in grape-juice. The objective of this study was to identify the effects of methionine on aroma compounds formation. Nitrogen source effects on growth, fermentative behaviour and aroma
Anelise Rodolfo Ferreira Pieralini et al.
The Journal of prosthetic dentistry, 108(1), 51-57 (2012-07-07)
Coatings of zirconite, Y(2)O(3) or ZrO(2) on wax patterns before investing in phosphate-bonded investments have been recommended to reduce the reaction layer in titanium castings, but they are not easily obtainable. Spinel-based investments are relatively stable with molten titanium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service