Skip to Content
Merck
All Photos(2)

Key Documents

V800040

Sigma-Aldrich

Ammonium cerium(IV) nitrate

AR, ≥98%

Synonym(s):

Ceric ammonium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NH4)2(NO3)6
CAS Number:
Molecular Weight:
548.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
AR
form:
crystalline powder

grade

AR

product line

Vetec

Assay

≥98%

form

crystalline powder

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Fe: ≤0.1%

SMILES string

N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.2HNO3.4NO3.2H3N/c;6*2-1(3)4;;/h;2*(H,2,3,4);;;;;2*1H3/q+4;;;4*-1;;

InChI key

WIBGOERAEYJBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit H Haviv et al.
Journal of the American Chemical Society, 132(36), 12519-12521 (2010-08-26)
The high-power ultrasonic irradiation of preformed magnetite (Fe(3)O(4)) nanoparticles in the presence of monoelectronic Ce-containing ceric ammonium nitrate [Ce(IV)(NH(4))(2)(NO(3))(6)] oxidant in MeCOMe afforded hydrophilic 50 nm-sized colloidal, highly stable maghemite (gamma-Fe(2)O(3)) nanoparticles. An "inorganic" Ce atom doping of the NP
Mohammad Mahdi Najafpour et al.
Dalton transactions (Cambridge, England : 2003), 41(34), 10292-10297 (2012-07-19)
According to UV-visible spectroscopy, X-ray diffraction spectrometry, dynamic light scattering, Fourier transform infrared spectroscopy, electron paramagnetic resonance spectroscopy, transmission electron microscopy, scanning electron microscopy and X-ray photoelectron spectroscopy, nano-sized manganese oxides are proposed as active catalysts for water oxidation in
Myron M Johnson et al.
The Journal of organic chemistry, 75(24), 8701-8704 (2010-11-23)
Reaction of (2,4,5-trimethoxyphenyl)(2-hydroxyphenyl)methanone with ceric ammonium nitrate furnished the xanthone, 2,3-dimethoxy-9H-xanthen-9-one. Under the same conditions the related (1,4-dimethoxynaphthalen-2-yl)(2-hydroxyphenyl)methanone resulted in the formation of 12a-methoxy-5H-benzo[c]xanthenes 5,7(12aH)-dione. Other examples of this novel transformation are also outlined.
Mazaahir Kidwai et al.
Chemical & pharmaceutical bulletin, 58(10), 1320-1323 (2010-10-12)
An environment friendly method for the synthesis of 2-oxo/thioxooctahydroquinazolin-5-one derivatives has been devised using Ceric ammonium nitrate (CAN) as catalyst and polyethylene glycol (PEG) as solvent. The cytotoxic effect of these compounds was studied on U87 human glioma cells, compounds
Kuppusamy Sujatha et al.
Bioorganic & medicinal chemistry letters, 19(15), 4501-4503 (2009-06-02)
An efficient and eco-friendly method for the synthesis of 4,4'-(arylmethylene)bis(1H-pyrazol-5-ols) has been accomplished by tandem Knoevenagel-Michael reaction of two equivalents of 5-methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one with various aromatic aldehydes catalyzed by ceric ammonium nitrate (CAN) in water. All the synthesized compounds 3a-j were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service