V800039
Ammonium carbonate
LR, 30-33%
Synonym(s):
Hartshorn salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein:
4253487
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
LR
form:
powder
Recommended Products
grade
LR
vapor density
2.7 (vs air)
product line
Vetec™
form
powder
concentration
30-33%
SMILES string
N.N.OC(O)=O
InChI
1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3
InChI key
PRKQVKDSMLBJBJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
N N Mal'tseva et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 62(3), 56-65 (2000-08-10)
The intensification of activity of associative and symbiotic nitrogen fixation while using low amounts of ammonium-carbonate compounds, carbonic-ammonium salts, in particular, has been established. The main ways and mechanisms of the action of ammonium-carbonate salts as a new and perspective
Debabrata Nandi et al.
Journal of colloid and interface science, 454, 69-79 (2015-05-26)
Reduced-graphene-oxide based superparamagnetic nanocomposite (GC) was fabricated and applied for the remediation of Ni(II) from an aqueous medium. The as-prepared GC was extensively characterized by Raman, TEM, AFM, SEM-EDX, SQUID, and BET analyses. Quantitative immobilization of Ni(II) in an aqueous
Tomohisa Hasunuma et al.
Biotechnology for biofuels, 7(1), 493-493 (2015-02-05)
To ensure reliable sources of energy and raw materials, the utilization of sustainable biomass has considerable advantages over petroleum-based energy sources. Photosynthetic algae have attracted attention as a third-generation feedstock for biofuel production, because algae cultivation does not directly compete
Itsuo Murakami et al.
PloS one, 9(12), e115359-e115359 (2014-12-17)
Resveratrol (RESV) is a plant polyphenol, which is thought to have beneficial metabolic effects in laboratory animals as well as in humans. Following oral administration, RESV is immediately catabolized, resulting in low bioavailability. This study compared RESV metabolites and their
Mirosława Różycka et al.
PloS one, 9(12), e114308-e114308 (2014-12-10)
Fish otoliths, biominerals composed of calcium carbonate with a small amount of organic matrix, are involved in the functioning of the inner ear. Starmaker (Stm) from zebrafish (Danio rerio) was the first protein found to be capable of controlling the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service