Skip to Content
Merck
All Photos(1)

Key Documents

V800004

Sigma-Aldrich

1,4-Dioxane

LR, ≥99%

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
LR
bp:
100-102 °C (lit.)
vapor pressure:
27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

grade

LR

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

product line

Vetec

Assay

≥99%

form

liquid

autoignition temp.

356 °F

expl. lim.

22 %

dilution

(for analytical testing)

refractive index

n20/D 1.422 (lit.)

pH

6-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

density

1.034 g/mL at 25 °C (lit.)

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huijiao Wang et al.
Journal of hazardous materials, 294, 90-98 (2015-04-12)
Degradation of 1,4-dioxane by ozonation, electrolysis, and their combined electro-peroxone (E-peroxone) process was investigated. The E-peroxone process used a carbon-polytetrafluorethylene cathode to electrocatalytically convert O2 in the sparged ozone generator effluent (O2 and O3 gas mixture) to H2O2. The electro-generated
Joshua J Pacheco et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(23), 8363-8367 (2014-06-10)
Terephthalic acid (PTA), a monomer in the synthesis of polyethylene terephthalate (PET), is obtained by the oxidation of petroleum-derived p-xylene. There is significant interest in the synthesis of renewable, biomass-derived PTA. Here, routes to PTA starting from oxidized products of
Daniel Gerrity et al.
Water research, 46(19), 6257-6272 (2012-10-16)
The performance of ozonation in wastewater depends on water quality and the ability to form hydroxyl radicals (·OH) to meet disinfection or contaminant transformation objectives. Since there are no on-line methods to assess ozone and ·OH exposure in wastewater, many
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)
Jonathan B Grimm et al.
Nature methods, 12(3), 244-250 (2015-01-20)
Specific labeling of biomolecules with bright fluorophores is the keystone of fluorescence microscopy. Genetically encoded self-labeling tag proteins can be coupled to synthetic dyes inside living cells, resulting in brighter reporters than fluorescent proteins. Intracellular labeling using these techniques requires

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service