Skip to Content
Merck
All Photos(1)

Key Documents

94026

Supelco

Amisulbrom Metabolite IT-14

PESTANAL®, analytical standard

Synonym(s):

3-Bromo-6-fluoro-2-methyl-1-((1-methyl-1H-1,2,4-triazol-3-yl)sulfonyl)-1H-indole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10BrFN4O2S
Molecular Weight:
373.20
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

suitability

suitable for LC-MS

format

neat

storage temp.

2-8°C

SMILES string

O=S(N1C(C)=C(Br)C2=C1C=C(F)C=C2)(C3=NN(C)C=N3)=O

Application

Environmental

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service