Skip to Content
Merck
All Photos(1)

Key Documents

819821

TOYOPEARL® LMW Size Exclusion Sampler, (HW-40, 50, 55F), 45 μm particles

LMW, phase hydroxylated methacrylic polymer, bottle of 3 × 150 mL, 45 μm particle size

Synonym(s):

TOYOPEARL® Size Exclusion Sampler

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500

product line

TOYOPEARL®

form

slurry

packaging

bottle of 3 × 150 mL

manufacturer/tradename

Tosoh 19821

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylic polymer phase

particle size

45 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TOYOPEARL® size exclusion media for gel filtration chromatography applications offer several important features:
  • High mechanical strength methacrylate for use at high flow rates
  • Stability in organic solvents
  • May be pressure packed at 5-7 bar
  • Wide pH range: 2-12
  • May be autoclaved

Packaged as a suspension in 20% aqueous ethanol.
TOYOPEARL media fractionate mixtures of proteins and other large molecular weight compounds over a wide size range (specific molecular weight ranges reported below were determined with globular proteins). Use these packings for such diverse analyses as separating RNA from protein, resolving oligosaccharides by degree of polymerization, and isolating agglutinin while maintaining a high hemagglutination titer.

These products are available in different particle size grades:
  • Superfine (20-40 μm): Highest efficiency and operating pressure; also used in thin layer gel filtration
  • Fine (30-60 μm): Recommended for lab preparative scale operations.
  • Medium (40-90 μm) and Coarse (50-100 μm): Used for large scale preparative chromatography where a high flow rate at a low pressure is essential.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

103.1 °F

Flash Point(C)

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service