Skip to Content
Merck
All Photos(1)

Key Documents

807129

TSKgel® ODS-120A (10 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, C18 (octadecyl) phase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

Product Name

TSKgel® ODS-120A HPLC Column, phase C18 (octadecyl), L × I.D. 30 cm × 7.8 mm, 10 μm particle size

material

stainless steel

Agency

suitable for USP L1

product line

TSKgel®

manufacturer/tradename

Tosoh 07129

extent of labeling

20% Carbon, multilayer, not encapped

parameter

0-100% organic solvent
0-45 °C temperature
3.0 mL/min flow rate
75 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

spherical silica

matrix active group

C18 (octadecyl) phase

particle size

10 μm

pore size

150 Å

operating pH

2.0-7.5

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel® ODS-120A columns contain 150Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel ODS-120A columns a good choice for the reversed phase chromatography of peptides, small proteins, and environmental samples such as poly-aromatic hydrocarbons. The "A" in the product description signifies that the material is not endcapped. For charged samples, the endcapped TSKgel ODS-120T column is a more suitable alternative.

Physical form

Shipped in 70% methanol - 30% water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service