Skip to Content
Merck
All Photos(1)

Key Documents

47303

Supelco

Isothermal Test Mix Kit

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813

grade

analytical standard

Quality Level

packaging

pkg of 3 × 2 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

General description

Isothermal Test Mix Kit comprises of three isothermal test mixes, used to indicate column efficiency, leaks, dead volume, and sample adsorption. Match the polarity of the test mix with the polarity of the column. This kit consists of 2 mL each of the three isothermal test mixes described below.
  • Nonpolar Column Test Mix - For all nonpolar phases. 500μg/mL each component in methylene chloride.
  • Intermediate Polarity Column Test Mix - For SPB-20, SPB-35, and other intermediate polarity phases. 500μg/mL each component in methylene chloride.
  • Polar Column Test Mix - For SUPELCOWAX 10, SP-1000, and other polar phases.
  • 500μg/mL each component in methylene chloride.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 47300-UNonpolar Column Test Mix, certified reference material, 500 μg/mL in dichloromethaneSDS

  • 47301Intermediate Polar Column Test Mix, certified reference material, 500 μg/mL in dichloromethaneSDS

  • 47302Polar Column Test Mix, certified reference material, 500 μg/mL each component in dichloromethaneSDS

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service