Skip to Content
Merck
All Photos(1)

Key Documents

13083-U

Supelco

Glass Packed GC Column (fits Agilent, Configuration "A")

phase 3% OV-1, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710

material

glass column

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

6.0 ft (1.8 m) × 1/4 in. × 2.0 mm

matrix

80/100 Chromosorb W HP support

matrix active group

3% OV-1 phase

column type

packed GC

compatibility

for use with Agilent Configuration A

Looking for similar products? Visit Product Comparison Guide

General description

In a packed column, the carrier gas percolates through the porous bed and separation is achieved by moving the solutes apart. Packed columns are made up of two main materials; stainless steel and Pyrex glass. Pyrex glass columns are shaped as desired by coiling at about 700°C. Glass column are in some cases treated with silanizing reagent to eliminate the surface hydroxyl groups which becomes catalytically active and produce asymmetric peaks. Chromosorb W is a fragile white solid prepared by heating the diatomaceous earth with a flux. The resulting fine structure is then cemented over glass which results in large and irregular pores. HP is the classification of a support as high performance grade.
Use a glass column for applications where high inertness is required, such as for the analysis of active analytes. All glass columns undergo a proprietary high temperature silanization to ensure inertness. This column fits Agilent 5880, 5890, 6890. 7820, and 7890 (configuration A, on-column injection, all detectors except TCD).

Application

Chromosorb W HP support packed glass GC colum was used in separating the Me3Si derivatives of the phenolic acids.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert L. Grob and Eugene F. Barry
Modern Practice of Gas Chromatography, 76-76 (2004)
Free, esterified, and insoluble-bound phenolic acids. 1. Extraction and purification procedure.
Krygier
Journal of Agricultural and Food Chemistry, 30, 330-334 (1982)
Ian A. Fowlis
Gas Chromatography, 27-27 (2008)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service