Skip to Content
Merck
All Photos(7)

Key Documents

ZRT1335

Sigma-Aldrich

Anti-HHV-8 LNA-1 Antibody, clone LN53 ZooMAb® Rat Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HHV-8/KSHV Orf73 LANA, HHV-8/KSHV Orf73 LNA, HHV-8/KSHV Orf73 LNA-1, HHV-8/KSHV Orf73 latency-associated nuclear antigen, HHV-8/KSHV Orf73 latent nuclear antigen, HHV-8/KSHV Orf73 latent nuclear antigen-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

LN53, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 119 kDa
observed mol wt ~80-105 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2cκ

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

vaccinia virus ... ORF73(4961527)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone LN53 is a ZooMAb® rat recombinant monoclonal antibody that specifically detects Human herpesvirus-8/Kaposi′s sarcoma-associated herpesvirus (HHV-8/KSHV) and macaque retroperitoneal fibromatosis herpesvirus (RFHV). It targets an epitope within the C-terminal domain.

Immunogen

Affinity-purified GST fusions of C-terminal fragments of HHV-8 LNA-1 (See: Kellam, P., et al. (1999). J. Virol. 73(6); 5149-5155).

Application

Quality Control Testing

Evaluated by Western Blotting in Body Cavity Based Lymphoma-1 (BCBL-1) cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected HHV-8 LNA-1 in Body Cavity Based Lymphoma-1 (BCBL-1) cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected HHV-8 LNA-1 in BCBL1 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected HHV-8 LNA-1 in Human Kaposi′s sarcoma tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Human herpesvirus-8 (also known as Kaposi′s sarcoma-associated herpesvirus, HHV-8, KSHV) is etiologically associated with all forms of Kaposi sarcoma (KS) including classic, endemic, transplant-related, and AIDS-related, as well as rare neoplastic disorders. It is a double stranded DNA virus with a large genome that encodes for over 87 open reading frames (ORFs) including genes necessary for capsid, tegument, envelope, DNA replication and regulatory proteins. HHV-8 undergoes a bi-phasic lifecycle and can persist indefinitely in the infected host in a latent form during which time only a small fraction of regulatory viral proteins is expressed. The most prominent protein expressed in latently infected cells is the ORF73 gene product known as the latency-associated nuclear antigen (LANA) that functions to ensure the maintenance of the viral genome by tethering the viral episomal DNA to host cell chromosomes. The ORF59 (also known as PF-8), a phosphoprotein, is reported to be a processivity factor for HHV-8 DNA polymerase (Pol-8). It interacts with and stimulates Pol-8 DNA synthesis activity. It is an early-late gene product localized to HHV-8-infected cell nuclei. PF-8 interacts with HHV-8 DNA polymerase (Pol-8) and stimulates Pol-8 DNA synthesis activity. Amino acid residues 10-27 and 279-301 are reported to be critical for the processivity function of PF-8. Residues 10 to 27 are essential for its interaction with Pol-8 and 279-301 are required for its binding to dsDNA. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Strahan, RC., et al. (2017). PLoS Pathog 13(7); e1006482; Chan, SR., and Chandran, B (2000). J. Virol. 74(23); 10920-10929; Kellam, P., et al. (1999). J. Virol. 73(6); 5149-5155; Chan, SR., et al. (1998). 240: 118-126).

Physical form

Purified recombinant rat monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service