Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2907

Anti-ZIP14 Antibody, clone 2J18 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, Metal cation symporter ZIP14, Solute carrier family 39 member 14, ZIP-14, Zrt- and Irt-like protein 14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 54.21 kDa
observed mol wt ~60 kDa

purified by

using Protein A

species reactivity

human, rat

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2J18 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects ZIP-14. It targets an epitope within 21 amino acids from the N-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the N-terminal, extracellular domain of human ZIP14.

Application

Quality Control Testing

Evaluated by Western Blotting in rat liver tissue lysate.

Western Blotting Analysis (WB): A 1:1,000 dilution from a representative lot detected ZIP14 in lysate from rat liver tissue.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ZIP14 in lysate from human heart tissue.

Affinity Binding Assay: A representative lot of this antibody bound with ZIP14 peptide with at least one thousand-fold (10,000X) higher affinity than with non-specific control peptide.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ZIP14 in human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ZIP14 in HepG2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Metal cation symporter ZIP14 (UniProt: Q15043; also known as Metal cation symporter ZIP14, LIV-1 subfamily of ZIP zinc transporter 4, LZT-Hs4, Solute carrier family 39 member 14, Zrt- and Irt-like protein 14, ZIP-14) is encoded by the SLC39A14 (also known as KIAA0062, ZIP14) gene (Gene ID: 23516) in human. The ZIP14 gene is mapped to human chromosome 8p21.3. The encoded protein is a transmembrane metal ion transporter and is a member of the LIV-1 subfamily. ZIP-14 is a multi-pass membrane protein and contains six transmembrane domains (TMDs), a metalloprotease, and a histidine-rich motif. It is predominantly expressed in the liver, pancreas, fetal liver, thyroid gland, heart and weakly expressed in spleen and thymus. ZIP-14 primarily functions as a divalent metal transporter specific for zinc (Zn). However, some studies report, it also mediates the transport of manganese (Mn), iron (Fe), and cadmium (Cd). As a zinc importing protein, ZIP-14 has an important role in glucose homeostasis. ZIP-14 along with ZIP8 are prime Mn2+ importers of the brain s capillary bed and in human brain microvascular endothelial cells (HBMVEC) in in-vitro conditions. Mutations in the ZIP14 gene lead to manganese dyshomeostasis, which in turn results in manganese accumulation in childhood-onset parkinsonism dystonia patients. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Maxel, T., et al. (2019). Scientific Reports. 9; 8589; McCabe, S., et al. (2023). Computational and Structural Biotechnology Journal. 21; 2332-2338).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service