Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2640

Sigma-Aldrich

Anti-TMEM106B Antibody, clone 2E4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Transmembrane protein 106B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 31.13 kDa
observed mol wt ~42 kDa

purified by

using Protein A

species reactivity

rat, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal cytoplasmic domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

Related Categories

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2E4 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Transmembrane protein 106B (TMEM106B). It targets an epitope within 22 amino acids from the N-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the N-terminal, cytoplasmic domain of human Transmembrane protein 106B (TMEM106B).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TMEM106B in Jurkat cell lysate.

Tested Applications

Affinity Binding Assay: A representative lot of this antibody bound TMEM106B peptide with a KD of 1.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TMEM106B in Human testis and Rat brain tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TMEM106B in U2OS cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Transmembrane protein 106B (UniProt: Q9NUM4; also known as TMEM106B) is encoded by the TMEM106B gene (Gene ID: 54664) in human. TMEM106B is a single-pass type II membrane protein that contains a cytoplasmic domain (aa 2-96), a transmembrane domain (aa 97-117), and a lumenal domain (aa 118-274). Its expression is observed in the brain, including the frontal cortex. Some expression has also been reported in lung epithelial cells. In neurons, it is involved in the transport of late endosomes/lysosomes and may also be involved in dendrite morphogenesis and maintenance by regulating lysosomal trafficking. It may serve as a molecular brake for retrograde transport of late endosomes/lysosomes, possibly via its interaction with MAP6. It also plays a role in the regulation of lysosomal size and responsiveness to stress and is required for lysosomal acidification. A truncated C-terminal fragment of TMEM106B (aa 120-354) has also been reported that aggregates into stable amyloid fibrils in the brain of subjects suffering from diverse genetic and sporadic tauopathies, amyloid-β amyloidoses, and synucleinopathies. It also plays a role in ACE2-independent SARS-CoV-2 entry into cell where it is required for post-endocytic stage of virus entry by facilitating spike-mediated membrane fusion. Mutations in TMEM106B gene have been linked to Ubiquitin-positive frontotemporal dementia (UP-FTD), an autosomal dominant neurodegenerative disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Baggen, J., et al. (2023). Cell. 186(16); 3427-3442; Stagi, M., et al. (2014). Mol. Cell Neurosci. 61: 226-240).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service