Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2500

Sigma-Aldrich

Anti-AMIGO-2 Antibody, clone 1B9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ALI1, AMIGO-2, Alivin-1, Differentially expressed in gastric adenocarcinomas (DEGA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B9, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58 kDa
observed mol wt ~70 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... AMIGO2(347902)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1B9 is a ZooMAb® rabbit recombinant monoclonal antibody that detects AMIGO-2. It targets an epitope within 26 amino acids from the C-terminal extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 26 amino acids from the C-terminal extracellular domain of human AMIGO-2.

Application

Quality Control Testing

Evaluated by Western Blotting in K562 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AMIGO-2 in K562 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000-10,000 dilution from a representative lot detected AMIGO-2 in lysates from Y79, A549 cells and human kidney tissue.

Affinity Binding Assay: A representative lot of this antibody bound AMIGO-2 peptide with at least one hundred thousand-fold (100,000X) higher than with non-specific control peptide.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected AMIGO-2 in K562 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected AMIGO-2 in human stomach tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Amphoterin-induced protein 2 (UniProt: Q86SJ2; also known as AMIGO-2, Alivin-1, Differentially expressed in gastric adenocarcinomas (DEGA)) is encoded by the AMIGO2 (also known as ALI1) gene (Gene ID: 347902) in human. AMIGO-2 is a member of the AMIGO family, which includes three, type I transmembrane proteins: AMIGO-1, AMIGO-2, and AMIGO-3. These proteins are known to play a role in nervous system development and have been implicated in various neuronal activities. AMIGO-2 protein contains 6 leucine-rich repeats (LRR), an Ig-like C2-type domain in the extracellular region, a transmembrane domain (aa 399-419), and a cytoplasmic domain (aa 420-522). It is involved in cell adhesion, axon tract development, and synaptic formation within the nervous system. The AMIGO family of proteins exhibit both homophilic and heterophilic binding activity. AMIGO-2 is primarily expressed in the nervous system. Its expression is typically higher during the early stages of brain development. AMIGO-2 have been linked to several neurological, neurodevelopmental disorders and can influence neural regeneration processes. Changes in AMIGO-2 expression levels have been observed in certain cancers and it is differentially expressed in human gastric adenocarcinomas. (Ref.: Chen, Y et al., (2006), Brain Research Reviews, 51, 265 274).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service