Skip to Content
Merck
All Photos(11)

Key Documents

ZRB2104

Sigma-Aldrich

Anti-TRAF2 Antibody, clone 3F11 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

TNF receptor-associated factor 2;EC:2.3.2.27;E3 ubiquitin-protein ligase TRAF2;RING-type E3 ubiquitin transferase TRAF2;Tumor necrosis factor type 2 receptor-associated protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F11, recombinant monoclonal

description

3F11 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 55.86 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

mouse, canine, porcine, horse, monkey, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... TRAF2(7186)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3F11 is a ZooMAb® Mouse recombinant monoclonal antibody that specifically detects TNF receptor-associated factor 2 (TRAF2). It targets an epitope within 15 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human TNF receptor-associated factor 2 (TRAF2).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected TRAF2 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TRAF2 in Jurkat cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TRAF2 in human kidney tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound TRAF2 peptide with a KD of 4.2 x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TRAF2 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

TNF receptor-associated factor 2 (UniProt: Q12933; also known as EC:2.3.2.27, E3 ubiquitin-protein ligase TRAF2, RING-type E3 ubiquitin transferase TRAF2, Tumor necrosis factor type 2 receptor-associated protein 3) is encoded by the TRAF2 (also known as TRAP3) gene (Gene ID: 7186) in human. TRAF2 is homodimeric protein that regulated the activation of  NF-kB and JNK and plays a key role in the regulation of cell survival and apoptosis. It can also form heterotrimer with TRAF1 or TRAF3. It is required for normal antibody isotype switching from IgM to IgG. It exhibits E3 ubiquitin-protein ligase activity and is reported to promote lysine 63-linked ubiquitination of target proteins, such as BIRC3, RIPK1 and TICAM1. It regulates BIRC2 and BIRC3 protein levels by inhibiting their autoubiquitination and subsequent degradation. It is an essential constituent of several E3 ubiquitin-protein ligase complexes, where it promotes the ubiquitination of target proteins by bringing them into contact with other E3 ubiquitin ligases. However, in the absence of sphingosine-1-phosphate it displays very low E3 ubiquitin ligase activity. TRAF2 has a RING-type zinc finger domain (aa 34-73) that is essential for its E3 ubiquitin ligase activity. Its coiled-coil domain (aa 299-348) mediate it homo- and hetero-oligomerization. It has two TRAF-type zinc finger domains (aa 124-180 and 177-233).  It also contains a MATH domain (aa 351-496) that binds to receptor cytoplasmic domains. TRAF2 undergoes phosphorylation at several serine residues within the first 128 amino acids. Its phosphorylation at threonine 117 is required for lysine 63- linked polyubiquitination and is also essential for its interaction with IKKα and IKKβ and subsequent activation of NF-κB. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Zhang, L., et al. (2010). J. Mol. Biol. 396(3); 528-539; Alvarez, SE., et al. (2010). Nature. 465(7301); 1084-1088; Gil, J., et al. (2004). Mol. Cell Biol. 24(10); 4502-4512).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service