Skip to Content
Merck
All Photos(6)

Key Documents

ZMS1213

Sigma-Aldrich

Anti-Phosphoepitope SR Antibody, clone 1H4 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

SR, Ser-Arg-Rich

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

Clone, 1H4

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~55, kDa
observed mol wt ~30, kDa

purified by

using protein G

species reactivity

rat, human

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1H4 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Ser-Arg rich (SR) phosphoproteins.

Immunogen

Nuclei from the oocytes of Xenopus laevis.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from L6 cells treated with insulin.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Phosphoepitope SR proteins in lysates from L6 cells treated with insulin (100 nM; 5 min) but not in untreated cells.

Tested Applications

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Phosphoepitope SR proteins in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Ser-Arg-rich (SR) proteins make up a family of functionally and structurally conserved phosphoproteins that are required for alternative and constitutive pre-mRNA splicing. These proteins are involved in various aspects of pre-mRNA splicing and in spliceosome assembly including the identification and appropriation of splice-sites, and the manipulation of alternative splicing regulation. During mitosis, SR proteins are diffusely distributed in the cytosol, but later in mitosis, nuclear speckles are assembled into cytoplasmic structures known as mitotic interchromatin granules that disappear around telophase, concomitant with the reconstruction of nuclear speckles just after nuclear envelope/lamina formation. These proteins are characterized by their modular composition consisting of two domains of interest: an N-terminal RNA recognition motif (RRM) that serves in the determination of DNA-binding specificity, and the RS domain, an arginine and serine rich, C-terminal domain that serves as a shuttling and localization director of SR proteins and as a splicing activation domain. The RS domains are extensively phosphorylated, and this phosphorylation is essential for nuclear import of SR proteins and for their functions in mediating spliceosome assembly. Partial dephosphorylation of SR proteins is also critical for progression of the assembled spliceosome to catalysis and for a series of post-splicing events from the interaction with the mRNA transport machinery to SR protein-mediated translational control. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Saitoh, M., et al. (2012). Mol. Biol. Cell. 15; 23(6): 1115-1128; Zhong, XY., et al. (2009). Genes Dev. 23(4); 482-495; Prasanth, KV., et al. (2003). Mol. Biol. Cell. 14(3); 1043-1057).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service