Z744289
Wilmad® 3.2 mm ZrO2 Open Ended Rotor for Bruker® MAS Probe
zirconia, pack of 1 ea
Synonym(s):
Solid State NMR, MAS NMR,
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23152100
NACRES:
NB.15
Recommended Products
material
zirconia
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WP-501-3180
General description
Manufactured from the highest purity Zirconia, our MAS Rotors provide the NMR spectroscopist with the ultimate alternative for analysis of solid samples; the need to solvate is eliminated. They are available for most present solid state NMR spectrometers. Wilmad′s MAS Rotors are carefully examined for material irregularities by optical methods before and after the precision machining process. Without over-spinning, each rotor is spin tested to only the highest specified spinning speed. Overspin testing, a common practice with some rotor brands, can actually damage the rotor prior to use. High precision (which becomes a necessity for proper spinning performance) is maintained in the manufacture of the end caps*. Most caps are fitted with O-rings for better sealing; some have axial holes for venting. Advantages of Zirconia Rotors Most homogeneous ceramic material structure. Wilmad′s Zirconia Rotor Body has higher strength (1,000 MPa, greater than Si3N4). Highest spinning speed High precision superior finish (0.05 µ mirror gloss). Some of our caps have a uniquely designed detachable O-ring. Use it when a gas-tight environment is required for the sample.• Application Temperature:-150 to 650 Degree C• Vmax =24 kHz
Application
NMR
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service