Z743613
Color coded Tough-Tags™, tube and spot labels
white to green, L × W × diam. 1.25 in. × 0.5625 in. × 0.44 in., roll of 500
Synonym(s):
solvent resistant labels, temperature resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Recommended Products
packaging
roll of 500
Quality Level
manufacturer/tradename
Diversified Biotech COLG-1000
L × W × diam.
1.25 in. × 0.5625 in. × 0.44 in.
color
white to green
Looking for similar products? Visit Product Comparison Guide
General description
COLOR-CODED TOUGH-TAGS
- Easily differentiate samples and identify what you need
- Translucent writing area keeps contents visible
- Repositionable for up to thirty minutes
- Adhesive strengthens over time
Legal Information
Tough-Tags is a trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service