SRP3281
SCD30 LIGAND human
recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Synonym(s):
CD153, CD30L, TNFSF8
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.77
Recommended Products
biological source
human
recombinant
expressed in CHO cells
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
potency
10.0-25.0 ng/mL
mol wt
30.0-50.0 kDa
packaging
pkg of 50 μg
impurities
<0.1 EU/μg endotoxin, tested
color
white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TNFSF8(944)
General description
CD30L binds specifically to CD30 (receptor), which is expressed on activated, but not resting, B and T cells, in lymphomas and various chronically inflamed tissues. CD30L/CD30 interactions initiate a signaling cascade that can ultimately lead to the activation of NF-KappaB. CD30L/CD30 signaling exerts pleiotropic effects on normal cells, including cell death, differentiation, or cell division. Certain diseases, including Hodgkins′s lymphoma, allergic inflammation, diabetes (in NOD mice), and mycobacterial infection can also be affected by CD30L/CD30 signaling. The CD30L gene encodes for a 234 amino acid type II transmembrane protein, which contains a 37 amino acid cytoplasmic sequence, a 25 amino acid transmembrane domain and a 172 amino acid extracellular domain.
Biochem/physiol Actions
CD30 ligand (CD30L) is a type-II membrane-associated glycoprotein protein belonging to the TNF superfamily and is expressed primarily on certain B cells, T cells, and monocytes. Recombinant human soluble CD30L (sCD30L) is a 188 amino acid polypeptide corresponding to the extracellular domain, and contains an 8 residue N-terminal His -Tag.
Physical form
Lyophilized from 1x PBS.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mary K Kennedy et al.
Immunology, 118(2), 143-152 (2006-06-15)
Ligands and receptors in the tumour necrosis factor (TNF) and tumour necrosis factor receptor (TNFR) superfamilies have been the subject of extensive investigation over the past 10-15 years. For certain TNFR family members, such as Fas and CD40, some of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service