Skip to Content
Merck
All Photos(1)

Key Documents

SRP3002

Sigma-Aldrich

4-1BBL human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CD137L, TNFSF9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

5-10 ng/mL ED50

mol wt

19.5 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFSF9(8744)

General description

4-1BBL, a member of the TNF superfamily, is expressed in B cells, dendritic cells, activated T cells and macrophages. 4-1BBL binds to its receptor 4-1BB, and provides a co-stimulatory signal for T cell activation and expansion. The human 4-1BBL gene codes for a 254 amino acid type II transmembrane containing a 28 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 205 amino acid extracellular domain. The soluble form of 4-1BL contains the TNF-like portion of the extracellular domain of 4-1BBL. Recombinant human 4-1BBL is a soluble 19.5 kDa protein consisting of 185 amino acid residues.

Biochem/physiol Actions

4-1BBL, a member of the TNF superfamily, is expressed in B cells, dendritic cells, activated T cells and macrophages. The soluble form of 4-1BBL contains the TNF-like portion of the extracellular domain of 4-1BBL. Recombinant human 4-1BBL is a soluble 19.5 kDa protein consisting of 185 amino acid residues.

Sequence

MREGPELSPD DPAGLLDLRQ GMFAQLVAQN VLLIDGPLSW YSDPGLAGVS LTGGLSYKED TKELVVAKAG VYYVFFQLEL RRVVAGEGSG SVSLALHLQP LRSAAGAAAL ALTVDLPPAS SEARNSAFGF QGRLLHLSAG QRLGVHLHTE ARARHAWQLT QGATVLGLFR VTPEIPAGLP SPRSE

Physical form

Lyophilized from 5 mM Tris, pH 8.0 + 75 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Alderson et al.
European journal of immunology, 24(9), 2219-2227 (1994-09-01)
4-1BB was originally described as a cDNA expressed by activated murine T cells and subsequently demonstrated to encode a member of the tumor necrosis factor receptor family of integral membrane proteins. Recently, we identified and cloned a murine ligand for
D S Vinay et al.
Seminars in immunology, 10(6), 481-489 (1998-11-25)
4-1BB is an inducible T cell surface receptor which belongs to the tumor necrosis factor receptor superfamily, a group of cysteine-rich cell-surface molecules. Both human and mouse 4-1BB recently received HLDA nomenclature. Naive T cells lack 4-1BB, which is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service