Skip to Content
Merck
All Photos(2)

Key Documents

SRP0735

Sigma-Aldrich

LIMK2, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

2.5-3.5 nmol/min·mg (radiometric)

mol wt

~96 kDa

concentration

0.05 μg/μL

accession no.

CR456513

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LIMK2(3985)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant full-length human LIMK2 was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. LIMK2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain (1). LIM domains are highly conserved cysteine-rich structures that can mediate protein-protein interaction. LIMK2 is phosphorylated and activated by ROCK, a downstream effector of Rho, and then LIMK2 in turn phosphorylates cofilin, inhibiting its actin-depolymerizing activity which contributes to Rho-induced reorganization of the actin cytoskeleton (2). LIMK2 is highly expressed in liver, colon, stomach, spleen, brain, kidney, and placenta.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service