Skip to Content
Merck
All Photos(2)

Key Documents

SRP0713

Sigma-Aldrich

PFTK1 (CDK14)/CYCLINY, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

2.0-3.5 nmol/min·mg (radiometric)

mol wt

~64 kDa (Cyclin Y)
~72 kDa (PFTK1 (CDK14))

concentration

0.1 μg/μL

accession no.

NM_012395
NM_145012

shipped in

dry ice

storage temp.

−70°C

Gene Information

Application

Useful for Kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human full-length PFTK1 (CDK14) and Cyclin Y (2-end) were co-expressed by Baculovirus in Sf9 insect cells using N-terminal GST tags. PFTK1 is a member of the CDC2 - related protein kinase family which is expressed primarily in the postnatal and adult nervous system (1). PFTK1 is expressed at high levels in brain, pancreas, kidney, heart, testis, and ovary. PFTK1 amino acid residue ser119 is required for its interaction with all four 14-3-3 isoforms, while ser120 may contribute slightly to PFTK1 binding (2). PFTK1 acts as a cyclin-dependent kinase that regulates cell cycle progression and cell proliferation. Overexpression of PFTK1 predicts resistance to chemotherapy in patients with oesophageal squamous cell carcinoma.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service