recombinant
expressed in baculovirus infected Sf9 cells
Assay
70-95% (SDS-PAGE)
form
frozen liquid
specific activity
0.2-1.0 nmol/min·mg (radiometric)
mol wt
~82 kDa
concentration
0.1 μg/μL
accession no.
NM_005012
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... ROR1(4919)
Biochem/physiol Actions
Recombinant human ROR1(429-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. ROR1 is a glycosylated type I membrane protein that belongs to the ROR subfamily of cell surface receptors that play a role in the control of most basic cellular processes including cell proliferation, differentiation, migration and metabolism (1). Many human breast cancers express the ROR1 protein and high-level expression of ROR1 in breast adenocarcinoma is associated with aggressive disease. Silencing expression of ROR1 in human breast cancer cell lines found to express this protein impaired their growth in vitro and also in immune-deficient mice (2).
Storage and Stability
Store in aliquots at -70°C. Avoid freeze/thaw cycles.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service