Skip to Content
Merck
All Photos(1)

Key Documents

SRP0285

Sigma-Aldrich

Sphingosine kinase 2 Active human

recombinant, expressed in baculovirus, ≥80% (SDS-PAGE)

Synonym(s):

SK2, SPHK2, SPK 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.77

recombinant

expressed in baculovirus

Assay

≥80% (SDS-PAGE)

form

aqueous solution

specific activity

≥20 units/μg

mol wt

69.5 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SPHK2(56848)

General description

Human Sphingosine kinase 2 (GenBank Accession No. AF245447), full length with N-terminal His tag, MW = 69.5 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Sphingosine kinase 2 is useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling. Sphingosine kinase 2 has been used in a study to assess the expression of sphingosine kinase 1 in oestrogen receptor-negative breast cancer. Sphingosine kinase 2 has also been used in a study to investigate the antitumor effects of sorafenib (≤ 10 μM) on four human neuroblastoma cell lines.

Unit Definition

One unit of sphingosine kinase activity is defined as the amount of enzyme required to produce 1 pmol of S1P/min

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gemma Fabrias et al.
Progress in lipid research, 51(2), 82-94 (2011-12-28)
Sphingolipids are a wide family of lipids that share common sphingoid backbones, including (2S,3R)-2-amino-4-octadecane-1,3-diol (dihydrosphingosine) and (2S,3R,4E)-2-amino-4-octadecene-1,3-diol (sphingosine). The metabolism and biological functions of sphingolipids derived from sphingosine have been the subject of many reviews. In contrast, dihydrosphingolipids have received
Fan Yang et al.
Cancer biology & therapy, 13(7), 534-541 (2012-03-13)
Neuroblastoma is the most common extracranial solid tumor in the pediatric population. Sorafenib (Nexavar), a multikinase inhibitor, blocks cell proliferation and induces apoptosis in certain types of cancers. Here, we tested antitumor effects of sorafenib (≤ 10 µM) on four
J Ohotski et al.
British journal of cancer, 106(8), 1453-1459 (2012-03-31)
We previously reported that sphingosine 1-phosphate receptor 4 (S1P(4)) is expressed and stimulates the ERK-1/2 pathway via a human epidermal growth factor receptor 2 (HER2)-dependent mechanism in oestrogen receptor-negative (ER(-)) MDA-MB-453 breast cancer cells. Clinical relevance of S1P(4) and sphingosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service