Skip to Content
Merck
All Photos(1)

Key Documents

SML4119

Fisetin

new

≥95% (HPLC)

Synonym(s):

2-(3,4-Dihydroxyphenyl)-3,7-dihydroxy-4H-1-benzopyran-4-one, 3,3′,4′,7-Tetrahydroxyflavone; 5-Desoxyquercetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6
CAS Number:
Molecular Weight:
286.24
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

Neuroprotective and antioxidant, slow cognitive decline in mice models of neurodegeneration.


Fisetin is a natural flavonoid found in various fruits and vegetables that exhibits neuroprotective and antioxidant activities. It inhibits PI 3-K, Akt, mTOR and Cdk6. Fisetin slow cognitive decline and the spread of pathology in mouse models of neurodegeneration.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service