Skip to Content
Merck
All Photos(1)

Key Documents

SML4085

Pasireotide Acetate

new

≥95% (HPLC)

Synonym(s):

Cyclo[(2S)-2-phenylglycyl-D-tryptophyl-L-lysyl-O-(phenylmethyl)-L-tyrosyl-L-phenylalanyl-(4R)-4-[[[(2-aminoethyl)amino]carbonyl]oxy]-L-prolyl] acetate, Cyclo[Phg-DTrp-Lys-Tyr(Bzl)-Phe-(2-aminoethy-carbamoyloxy-Pro)] acetate, SOM 230 acetate, SOM-230 acetate, SOM230 acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H66N10O9 · xC2H4O2
CAS Number:
Molecular Weight:
1047.21 (free base basis)
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Somatostatin (SRIF) eptidomimetic with high affinity for somatostatin receptor subtypes STT1/2/3/5 and in vivo efficacy for suppressing GH secretion.
Pasireotide (SOM230) is a somatostatin (somatotropin release inhibiting factor, SRIF) eptidomimetic with high affinity for somatostatin receptor subtypes STT1/2/3/5 (IC50 = 9.3/1/1.5/0.16 nM), but not STT4 (IC50 > 1 μM). SOM230 potently inhibits the growth hormone (GH) releasing hormone (GHRH)-induced GH release in primary rat pituitary cultures (IC50 = 0:4 nM) and suppresses the secretion of GH, insulin and glucagon in rats in vivo (ED50 in μg/kg post 1h administration via s.c. = 0.22, 3.4, 74, respectively; GH ED50 post 6h = 5.5 μg/kg).

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service