Skip to Content
Merck
All Photos(1)

Key Documents

SML4079

BI-3802

new

≥98% (HPLC)

Synonym(s):

2‐[(6‐{[5‐Chloro‐2‐(cis-3,5‐dimethylpiperidin‐1‐yl)pyrimidin‐4‐yl]amino}‐1-methyl‐2‐oxo‐1,2‐dihydroquinolin‐3‐yl)oxy]‐N-methylacetamide, BI 3802, BI3802

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H29ClN6O3
CAS Number:
Molecular Weight:
484.98
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent and selective BCL6 degrader that targets BTB domain with high-affinity.
BI-3802 is a potent and selective BCL6 degrader that targets BTB domain with high-affinity (IC50 ≤3 nM by ULight assay). BI-3802 induces BCL6 polymerization (filaments formation) and SIAH1 E3 ligase-mediated ubiquitination, leading to proteasomal degradation (by 80% post 90 min 5 µM treatment in SU-DHL-4 cells) and inhibition of BCL6-dependent proliferation in a dose-depenent manner (typical dosing range 0.04 - 3 µM).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service