SML4057
PF-9363
≥98% (HPLC)
Synonym(s):
2,6-Dimethoxy-N-[4-methoxy-6-(1H-pyrazol-1-ylmethyl)-1,2-benzisoxazol-3-yl]benzenesulfonamide, CTx 648, CTx-648, CTx648, PF9363
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C20H20N4O6S
CAS Number:
Molecular Weight:
444.46
MDL number:
UNSPSC Code:
41106305
UNSPSC Code:
12352200
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
Orally active, highly potent and selective lysine acetyltransferase KAT6A inhibitor with anti-cancer efficacy in vivo.PF-9363 (CTx-648) is an orally active, highly potent and selective lysine acetyltransferase KAT6A inhibitor (KAT6A Ki = 0.27 nM; KAT6B/7/5/8 Ki = 2.4/70/420/670 nM). PF-9363 selectively inhibits H3K23 acetylation (ZR-75-1 H3K23Ac/H3K14Ac IC50 = 0.85/120 nM) and exhibits anti-proliferation potency against high KAT6A-expressing estrogen receptor signaling-positive (ER+) breast cancer cells in cultures (ZR-75-1 IC50 = 0.37 nM) and in mice in vivo (complete inhibition of ZR-75-1 xenograft tumor growth by 0.03 mg/kg/day p.o.) by downregulating cancr ER signaling, cell cycle and MYC transcriptional programs.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service