Skip to Content
Merck
All Photos(1)

Key Documents

SML4035

CGP 78608 hydrochloride

new

≥97%

Synonym(s):

CGP 78608 hydrochloride, CGP-78608 hydrochloride, CGP78608 hydrochloride, PAMQX hydrochloride, [(1S)-1-[[(7-Bromo-1,2,3,4-tetrahydro-2,3-dioxo-5-quinoxalinyl)methyl]amino]ethyl]phosphonate, hydrochloride, P-[(1S)-1-[[(7-Bromo-1,2,3,4-tetrahydro-2,3-dioxo-5-quinoxalinyl)methyl]amino]ethyl]phosphonic acid, hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13BrN3O5P·HCl
CAS Number:
Molecular Weight:
414.58
UNSPSC Code:
12352200

Quality Level

Assay

≥97%

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Potent glycine site-selective NMDA receptor antagonist with anticonvulsant efficacy in vivo.
CGP 78608 (PAMQX) is a potent glycine site-selective NMDA receptor antagonist (IC50 = 5 nM by competitive binding against 5 nM [3H]MDL 105,519; IC50 = 3/2.7 μM against AMPA/kainate, >10 μM against CGP39653 (glutamate site) and channel blocker MK-801) that exhibits anticonvulsant efficacy in mice in vivo (electroshock-induced convulsion ED50 = 12 mg/kg i.p.).

Caution

Hygroscopic

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service