Skip to Content
Merck
All Photos(1)

Key Documents

SML3992

Rhod-5N potassium salt

new

≥95% (HPLC)

Synonym(s):

9-[4-[bis(Carboxymethyl)amino]-3-[2-[2-[bis(carboxymethyl)amino]-5-nitrophenoxy]ethoxy]phenyl]-3,6-bis(dimethylamino)-xanthylium, potassium salt, Rhod 5N potassium salt, Rhod5N potassium salt, Rhodamine-5N potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H39N5O13
Molecular Weight:
785.75
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

, faint red to black

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(O)CN(C1=C(C=C(C=C1)C2=C3C=CC(N(C)C)=CC3=[O+]C4=C2C=CC(N(C)C)=C4)OCCOC5=CC([N+]([O-])=O)=CC=C5N(CC(O)=O)CC([O-])=O)CC(O)=O.[K]

Biochem/physiol Actions

Low-affinity fluorescent Ca2+ probe for measuring relatively high [Ca2+].
Rhod-5N is a low-affinity fluorescent Ca2+ probe (Kd = 320 μM) that is preferred over high-affinity probes, such as Rhod-2 (Kd = 570 nM), for measuring [Ca2+] when very little buffering can be tolerated or in subcellular compartments where relatively high Ca2+ levels are expected, such as mitochondria (MT) and endoplasmic reticulum (ER). Ex/Em = 551/576 nm.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service