Skip to Content
Merck
All Photos(2)

Key Documents

SML3911

Sigma-Aldrich

TCH-165

≥98% (HPLC)

Synonym(s):

Ethyl (4R,5R)-rel-1-benzyl-5-(4-(benzylamino)phenyl)-2-(4-methoxyphenyl)-4-phenyl-4,5-dihydro-1H-imidazole-4-carboxylate, rel-Ethyl (4R,5R)-4,5-dihydro-2-(4-methoxyphenyl)-4-phenyl-1-(phenylmethyl)-5-[4-[(phenylmethyl)amino]phenyl]-1H-imidazole-4-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H37N3O3
CAS Number:
Molecular Weight:
595.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N2([C@@H]([C@](N=C2c6ccc(cc6)OC)(c5ccccc5)C(=O)OCC)c3ccc(cc3)NCc4ccccc4)Cc1ccccc1

InChI

1S/C39H37N3O3/c1-3-45-38(43)39(33-17-11-6-12-18-33)36(31-19-23-34(24-20-31)40-27-29-13-7-4-8-14-29)42(28-30-15-9-5-10-16-30)37(41-39)32-21-25-35(44-2)26-22-32/h4-26,36,40H,3,27-28H2,1-2H3/t36-,39-/m1/s1

InChI key

XXDLWRCUPASJGY-AEGYFVCZSA-N

Biochem/physiol Actions

Cell-permeable and potent activator of 20S proteasome assembly that and enhances 20S-mediated proteolysis.

TCH-165 is a cell-permeable and potent activator of 20S proteasome assembly that favors a proteolytically active, open-gate 20S proteasome subcomplex and enhances 20S-mediated proteolysis (nearly 10-fold ≤1.5 µM) to the detriment of the 26S proteasome, in a time- and dose-dependent manner. TCH-165 restores proteostasis and enhances degradation of intrinsically disordered proteins (IDPs) both in purified protein assays and in cell culture (α-synuclein, tau441, c-MYC, cFos, ornithine decarboxylase). TCH 165 degrades SNAP29 and syntaxin 17 and reduces autophagosome-lysosome fusion. TCH165 inhibits cancer cell proliferation, impedes in vivo tumor growth, and offers neuroprotection from distinct dipeptide repeat (DPR) protein toxicity and is well tolerated in vivo (in mice and in dogs) at therapeutic concentrations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service