Skip to Content
Merck
All Photos(2)

Key Documents

SML3899

Sigma-Aldrich

L-371,257

≥98% (HPLC)

Synonym(s):

1-[4-[(1-acetyl-4-piperidinyl)oxy]-2-methoxybenzoyl]-4-(2-oxo-2H-3,1-benzoxazin-1(4H)-yl)-piperidine, L 371257

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H33N3O6
CAS Number:
Molecular Weight:
507.58
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Highly potent and selective oxytocin receptor antagonist
L-371,257 is a highly potent and selective antagonist of human oxytocin receptor. L-371,257 potently inhibits oxytocin-induced contractions of the rat uterus in vitro and in vivo. L-371,257 exhibits good oral availability in rats. It exhibits good peripheral selectivity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service