SML3354
NRD167
≥98% (HPLC)
Synonym(s):
Ethyl-(5S,8S)-5-((1H-indol-3-yl)methyl)-8-(((benzyloxy)carbonyl)amino)-2-methyl-4,7-dioxo-14-thioxo-3,6,13,15-tetraazaoctadecan-18-oate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C34H46N6O6S
CAS Number:
Molecular Weight:
666.83
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
Biochem/physiol Actions
Ethylester prodrug of a potent and selective sirtuin 5 (SIRT5) inhibitor against SIRT5-dependent cancer proliferation and survival.
RD167 is the ethylester prodrug of a potent and selective sirtuin 5 (SIRT5) inhibitor (IC50 = 370 nM; <10% inhibition of SIRT1/2/3/6 at 10 μM). NRD167 exhibits anti-proliferation (IC50 = 5-8 μM) and apoptosis-induction efficacy (>80% at 5-8 μM) against SIRT5-dependent, but not SIRT5-independent cells. NRD167 selectively impairs proliferation of AML, but not normal hematological progenitors (6.25-12.5 μM; CD34+ cells). Pretreatment of AML patients-derived cells by NRD167 (12.5-25 μM for 72 hrs) prior to xenograft greatly prevents the survival of injected mice.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service