Skip to Content
Merck
All Photos(1)

Key Documents

SML3064

Sigma-Aldrich

Atopaxar hydrobromide

≥98% (HPLC)

Synonym(s):

1-(3-tert-Butyl-4-methoxy-5-morpholinophenyl)-2-(5,6-diethoxy-7-fluoro-1-imino-1,3-dihydro-2H-isoindol-2-yl)ethanone hydrobromide, 2-(5,6-Diethoxy-7-fluoro-1,3-dihydro-1-imino-2H-isoindol-2-yl)-1-[3-(1,1-dimethylethyl)-4-methoxy-5-(4-morpholinyl)phenyl]ethanone, monohydrobromide, E 5555, E-5555, E5555, ER 172594-06, ER-172594-00 hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H38FN3O5 · xHBr
CAS Number:
Molecular Weight:
527.63 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CN1C(C2=C(C(OCC)=C(C=C2C1)OCC)F)=N)C(C=C3N4CCOCC4)=CC(C(C)(C)C)=C3OC.[xHBr]

InChI

1S/C29H38FN3O5/c1-7-37-23-15-19-16-33(28(31)24(19)25(30)27(23)38-8-2)17-22(34)18-13-20(29(3,4)5)26(35-6)21(14-18)32-9-11-36-12-10-32/h13-15,31H,7-12,16-17H2,1-6H3

InChI key

QWKAUGRRIXBIPO-UHFFFAOYSA-N

Biochem/physiol Actions

Atopaxar (E5555) is an orally active, potent and selective thrombin receptor (protease-activated receptor; PAR1) antagonist (IC50 = 19 nM/E5555 vs. 490 nM/TRAP against 10 nM haTRAP for PAR-1 binding on human platelet membranes) that inhibits platelet aggregation induced by thrombin or TRAP T (human platelet IC50 = 64/31 nM against 1 u/mL Thrombin/5 μM TRAP; guinea pig IC50 = 13/97 nM against 2 u/mL Thrombin/20 μM TRAP), but not ADP, U46619, collagen, PAR-4ap or PAF. Atopaxar exhibits antithrombotic efficacy in guinea pigs and rats in vivo (10, 30, 100 mg/kg p.o.).
Orally active, potent and selective thrombin receptor (protease-activated receptor; PAR1) antagonist with antithrombotic efficacy in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service