Skip to Content
Merck
All Photos(1)

Key Documents

SML2902

Sigma-Aldrich

LP-922056

≥98% (HPLC)

Synonym(s):

2-((6-Chloro-7-cyclopropylthieno[3,2-d]pyrimidin-4-yl)thio)acetic acid, LP 922056

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9ClN2O2S2
CAS Number:
Molecular Weight:
300.78
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

[s]1c2c(ncnc2SCC(=O)O)c(c1Cl)C3CC3

InChI

1S/C11H9ClN2O2S2/c12-10-7(5-1-2-5)8-9(18-10)11(14-4-13-8)17-3-6(15)16/h4-5H,1-3H2,(H,15,16)

InChI key

LJYRIWUQISYYHA-UHFFFAOYSA-N

Biochem/physiol Actions

LP-922056 is an orally active, potent and selective NOTUM pectinacetylesterase inhibitor that activates Wnt signaling. LP-922056 stimulates endocortical bone formation in rodents and in humanized NOTUM mouse line.
orally active, potent and selective NOTUM pectinacetylesterase inhibitor that activates Wnt signaling and stimulates bone formation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service