SML2577
Pixantrone dimaleate
≥98% (HPLC)
Synonym(s):
2-Azaanthracene-9,10-dione dimaleate, 6,9-Bis[(2-aminoethyl)amino]benzo[g]isoquinoline-5,10-dione dimaleate, BBR 2778, BBR-2778, BBR2778, Pixantrone maleate
About This Item
Recommended Products
Assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
purple to dark blue
solubility
H2O: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
N(CCN)c1c2c(c(cc1)NCCN)C(=O)c3c(cncc3)C2=O.OC(=O)\C=C\C(=O)O.OC(=O)\C=C\C(=O)O
InChI
1S/C17H19N5O2.2C4H4O4/c18-4-7-21-12-1-2-13(22-8-5-19)15-14(12)16(23)10-3-6-20-9-11(10)17(15)24;2*5-3(6)1-2-4(7)8/h1-3,6,9,21-22H,4-5,7-8,18-19H2;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+
InChI key
SVAGFBGXEWPNJC-LVEZLNDCSA-N
Biochem/physiol Actions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Muta. 2 - Repr. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service