Skip to Content
Merck
All Photos(1)

Key Documents

SML2502

Sigma-Aldrich

MF-095

≥98% (HPLC)

Synonym(s):

(S)-N-(1-((5-(N-(tert-butyl)sulfamoyl)naphthalen-1-yl)amino)-1-oxo-3-phenylpropan-2-yl)cyclopropanecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N3O4S
CAS Number:
Molecular Weight:
493.62
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(NC(C)(C)C)c1c2c(c(ccc2)NC(=O)[C@@H](NC(=O)C4CC4)Cc3ccccc3)ccc1

InChI key

JSNVUWHDPLJARW-QHCPKHFHSA-N

Biochem/physiol Actions

Control probe for USP30 inhibitor MF-094
MF-095 is a control probe for MF-094, which is a potent inhibitor of ubiquitin specific protease 30 (USP30)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur F Kluge et al.
Bioorganic & medicinal chemistry letters, 28(15), 2655-2659 (2018-06-25)
Mitophagy is one of the processes that cells use to maintain overall health. An E3 ligase, parkin, ubiquitinates mitochondrial proteins prior to their degradation by autophagasomes. USP30 is an enzyme that de-ubiquitinates mitochondrial proteins; therefore, inhibiting this enzyme could foster

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service