Skip to Content
Merck
All Photos(1)

Key Documents

SML0275

Sigma-Aldrich

rac BHFF

≥98% (HPLC)

Synonym(s):

5,7-Bis(1,1-dimethylethyl)-3-hydroxy-3(trifluoromethyl) -2(3H)-benzofuranone, 5,7-Di-tert-butyl-3-hydroxy-3-trifluoromethyl-3H-benzofuran-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21F3O3
CAS Number:
Molecular Weight:
330.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: ≥15 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)(C)c1cc(c2OC(=O)C(O)(c2c1)C(F)(F)F)C(C)(C)C

InChI

1S/C17H21F3O3/c1-14(2,3)9-7-10(15(4,5)6)12-11(8-9)16(22,13(21)23-12)17(18,19)20/h7-8,22H,1-6H3

InChI key

RVNOANDLZIIFHB-UHFFFAOYSA-N

Biochem/physiol Actions

rac BHFF is a potent, selective GABA-B receptor positive allosteric modulator.
rac BHFF is an orally active, potent and selective positive allosteric modulator of GABA-B. rac BHFF increases the potency and efficacy of GABA.

Features and Benefits

This compound is featured on the GABAB Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to GABAB receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service