Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702119

Monoclonal Rat IgG2a Isotype Control, purified

new
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

RTG2A1-1, monoclonal

form

buffered aqueous solution

species reactivity

(This rat IgG2a kappa monoclonal antibody detects specifically the hemocyanin of keyhole limpet and can be used as a negative control antibody in other species.)

concentration

1 mg/ml

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This rat IgG2a kappa monoclonal antibody (clone RTG2A1-1, also known as G2A-1-1 or KLH/G2A-1-1) detects specifically the hemocyanin of keyhole limpet and can be used as a negative control antibody in other species.

Physical form

Phosphate buffered saline (PBS), pH 7.4, 15 mM sodium azide

Preparation Note

Purified by protein-G affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service