Skip to Content
Merck
All Photos(1)

Key Documents

SAB4702063

Monoclonal Anti-DLL4 purified low endotoxin produced in mouse

new

Synonym(s):

AOS6, Delta like ligand 4, canonical Notch ligand 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

conjugate

unconjugated

Quality Level

antibody product type

primary antibodies

clone

MHD4-46, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/ml

technique(s)

FUNC: suitable
flow cytometry: suitable

isotype

IgG1κ

UniProt accession no.

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... DLL4(54567)

General description

DLL4 (Delta-like 4) is one of five ligands of Notch receptors. It interacts with Notch1 and Notch4. DLL4 is up-regulated at sites of physiologic and pathologic angiogenesis, whereas its expression is low in most adult normal tissues. It is also highly expressed in human clear-cell renal carcinomas, bladder cancers, and breast cancers. Blocking the DLL4-Notch interaction seems to be a promissing therapeutic approach.

Specificity

The mouse monoclonal antibody MHD4-46 recognizes the extracellular domain of DLL4 (Delta-like ligand 4), a type I transmembrane protein which plays an important role in vascular development.

Immunogen

recombinant soluble human DLL4

Physical form

Phosphate buffered saline (PBS), pH 7.4

Preparation Note

Purified by protein-A affinity chromatography.

Storage and Stability

Store at 2-8°C. Do not freeze.

Other Notes

The antibody solution should be gently mixed before use.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service