Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503327

Sigma-Aldrich

Anti-USP42 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Deubiquitinating enzyme 42, USP42, Ubiquitin carboxyl-terminal hydrolase 42, Ubiquitin thioesterase 42, Ubiquitin-specific-processing protease 42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 145 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USP42(84132)

General description

Anti-USP42 Antibody detects endogenous levels of total USP42 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human USP42.

Immunogen Range: 251-300

Application

Anti-USP42 antibody produced in rabbit is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

USP42 (ubiquitin specific peptidase 42) is a nuclear protein belonging to the ubiquitin-specific protease (USP) family. It is widely distributed in human tissues, especially in skeletal muscle and testis. It is involved in the regulation of ubiquitin-mediated pathways. It exerts deubiquitylating activity by cleaving ubiquitin from an ubiquitin-β-galactosidase fusion protein. It plays an important role in the regulation of p53 activity. It helps to stabilize p53 in response to stress and promotes early activation of p53.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreas K Hock et al.
The EMBO journal, 30(24), 4921-4930 (2011-11-17)
The p53 tumour suppressor protein is a transcription factor that prevents oncogenic progression by activating the expression of apoptosis and cell-cycle arrest genes in stressed cells. The stability of p53 is tightly regulated by ubiquitin-dependent degradation, driven mainly by the
Nagore Elu et al.
Frontiers in physiology, 10, 534-534 (2019-05-28)
The ubiquitin E3 ligase UBE3A has been widely reported to interact with the proteasome, but it is still unclear how this enzyme regulates by ubiquitination the different proteasomal subunits. The proteasome receptor DDI1 has been identified both in Drosophila photoreceptor
Víctor Quesada et al.
Biochemical and biophysical research communications, 314(1), 54-62 (2004-01-13)
We have identified and cloned 22 human cDNAs encoding novel members of the ubiquitin-specific protease (USP) family. Eighteen of the identified proteins contain all structural features characteristic of these cysteine proteinases, whereas four of them have been classified as non-peptidase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service