Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502721

Sigma-Aldrich

Anti-Syndecan4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Amphiglycan, Ryudocan core protein, SDC4, SYND4, Syndecan-4 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SDC4(6385)

General description

The SDC-4 (syndecan-4) gene encodes a membrane-spanning proteoglycan which can bear heparan sulfate and chondroitin sulfate chains. Syndecan-4 is predominantly expressed on smooth muscle cells, epithelial cells and cardiomyocytes. The SDC-4 gene is mapped to human chromosome 20q13.12. Anti-Syndecan4 antibody detects endogenous levels of total syndecan-4 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Syndecan4.

Immunogen Range: 145-194

Biochem/physiol Actions

Syndecan-4 (SDC-4) is associated with bone development and osteogenesis by regulating fibroblast growth factor-2 (FGF2). SDC-4 gene expression promotes cell division, migration and adhesion. It is essential for wound healing and takes part in inflammation and fibrosis. An abnormal SDC-4 expression is observed in response to stress and metastatic tumor. Growth factors such as FGF2, TNF-α (tumor necrosis factor α) and TGF-β (transforming growth factor β) also tend to alter SDC-4 expression. It is implicated as a biomarker in cardiovascular disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Killing two birds with one stone: Dual blockade of integrin and FGF signaling through targeting syndecan-4 in postoperative capsular opacification
Qin Y, et al.
Cell Death & Disease, 8(7), e2920-e2920 (2017)
Regulation of vascular smooth muscle cell calcification by syndecan-4/FGF-2/PKCalpha signalling and cross-talk with TGFbeta
Borland SJ, et al.
Cardiovascular Research, 113(13), 1639-1652 (2017)
Lack of association between serum syndecan-4, myocardial fibrosis and ventricular dysfunction in subjects with chronic Chagas disease
Larocca TF, et al.
PLoS ONE, 12(12), e0189408-e0189408 (2017)
Chromosome 22q13. 3 deletion syndrome with a de novo interstitial 22q13. 3 cryptic deletion disrupting SHANK3
Delahaye A, et al.
European Journal of Medical Genetics, 52(5), 328-332 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service