Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500776

Sigma-Aldrich

Anti-EPHB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ELK, EPB1, EPHT2, EPTH2, Eph receptor B1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 109 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPHB1(2047)

General description

Anti-EPHB1 Antibody detects endogenous levels of total EPHB1 protein.
EPHB1(ephrin type-B receptor 1) belongs to the Eph gene family of receptor tyrosine kinases. It is present in venous endothelial cells during embryonic development to adulthood. It is located on human chromosome 3q22.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human EPHB1/2/3.

Immunogen Range: 631-680

Biochem/physiol Actions

EPHB1(ephrin type-B receptor 1) participates in carcinogenesis of RCC (renal cell carcinoma). It plays a major role in the growth of nervous and vascular system. It acts as a biomarker for squamous cell/adenosquamous carcinomas and adenocarcinomas of the gallbladder.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loss of expression of EphB1 protein in serous carcinoma of ovary associated with metastasis and poor survival
Wang H, et al.
International Journal of Clinical and Experimental Pathology (2013)
Single nucleotide polymorphism array analysis of uveal melanomas reveals that amplification of CNKSR3 is correlated with improved patient survival
Lake SL, et al.
The American Journal of Pathology (2013)
Decreased expression of receptor tyrosine kinase of EphB1 protein in renal cell carcinomas
Zhou S, et al.
International Journal of Clinical and Experimental Pathology, 7 (2014)
EphB1 and Ephrin-B, new potential biomarkers for squamous cell/adenosquamous carcinomas and adenocarcinomas of the gallbladder
Yuan Y, et al.
Asian Pacific Journal of Cancer Prevention, 15 (2014)
Beatriz Garcia-Diaz et al.
Acta neuropathologica, 138(3), 457-476 (2019-04-24)
Schwann cells (SC) enter the central nervous system (CNS) in pathophysiological conditions. However, how SC invade the CNS to remyelinate central axons remains undetermined. We studied SC migratory behavior ex vivo and in vivo after exogenous transplantation in the demyelinated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service