Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300285

Sigma-Aldrich

Anti-phospho-EGFR (pTyr1197) antibody produced in goat

affinity isolated antibody

Synonym(s):

Anti-ERBB antibody produced in goat, Anti-ERBB1 antibody produced in goat, Anti-HER1 antibody produced in goat, Anti-PIG61 antibody produced in goat, Anti-epidermal growth factor receptor (erythroblastic leukemia viral (v-erb-b) oncogene homolog, avian) antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~175 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(A-E-YP-L-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1197)

Gene Information

human ... EGFR(1956)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1197 (A-E-Y(p)-L-R)., according to the protein EGFR.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Receptor for EGF, but also for other members of the EGF family, as TGF-alpha, amphiregulin, betacellulin, heparin-binding EGF-like growth factor, GP30 and vaccinia virus growth factor. Is involved in the control of cell growth and differentia

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Javier Palazuelos et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(36), 11884-11896 (2014-09-05)
Several studies have elucidated the significance of a disintegrin and metalloproteinase proteins (ADAMs) in PNS myelination, but there is no evidence if they also play a role in oligodendrogenesis and CNS myelination. Our study identifies ADAM17, also called tumor necrosis
Lakshmi Reddy Bollu et al.
Cell cycle (Georgetown, Tex.), 13(15), 2415-2430 (2014-12-09)
Increased expressions of fatty acid synthase (FASN) and epidermal growth factor receptor (EGFR) are common in cancer cells. De novo synthesis of palmitate by FASN is critical for the survival of cancer cells via mechanisms independent of its role as
Yong Sang Hong et al.
PloS one, 9(10), e109440-e109440 (2014-10-29)
ERBB2-directed therapy is now a routine component of therapy for ERBB2-amplified metastatic gastroesophageal adenocarcinomas. However, there is little knowledge of the mechanisms by which these tumors develop acquired resistance to ERBB2 inhibition. To investigate this question we sought to characterize
X Chen et al.
Neoplasma, 61(5), 514-522 (2014-07-18)
Colorectal cancer (CRC) is the third common cancer and most of the chemotherapies of CRC currently used often suffer limited efficacy and large side effects. Targeted small-molecule by anti-tumor drugs are thought a promising strategy for improving the efficacy and reducing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service