Skip to Content
Merck
All Photos(1)

Key Documents

SAB3501162

Sigma-Aldrich

Monoclonal Anti-Hemagglutinin [4E11E1] antibody produced in mouse

affinity purified immunoglobulin (IgG)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin (IgG)

antibody product type

primary antibodies

clone

monoclonal

form

liquid

species reactivity

influenza A virus (avian | subtype H5N1)

concentration

1 mg/mL

technique(s)

western blot: suitable

isotype

IgG1
IgG2b

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Hemagglutinin Monoclonal Antibody: Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals, but it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. HA interacts with host cell surface proteins containing oligosaccharides with terminal sialyl residues. Its extracellular region has two domains (HA1 and HA2); HA1 is cleaved from the main hemagglutinin protein by the host immune system. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. This more recent virulent strain of H5N1 is now seen in Africa and Europe, as well as in Southeast Asia. There is some evidence of human to human spread of this virus, but it is thought that the efficiency of this type of transmission is low. Virus isolated from a human infected with the H5N1 strain in 1997 could bind to oligosaccharides from human as well as avian sources, indicating its species-jumping ability.

Immunogen

Recombinant protein corresponding to amino acids 17 - 338 of H5 HA1.

Application

Hemagglutinin antibody can be used for detection of H5 HA1 by Western blot at 2.5 μg/mL. Hemagglutinin antibody also specifically recognizes Avian H5N1 influenza virus at 2 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service